Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK11 SIRNA Assays ›
Gene Symbol:
NEK11
Gene Name:
NIMA related kinase 11
Gene Aliases:
-
Chromosome Location:
Chr.3: 131026850 - 131350465 on Build GRCh38
UniGene ID:
Hs.657336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36501
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146003.1
NP_001139475.1
7
916
NM_001321220.1
NP_001308149.1
7
916
NM_001321221.1
NP_001308150.1
9
1115
NM_001321223.1
NP_001308152.1
7
1034
NM_001321224.1
NP_001308153.1
6
750
NM_024800.4
NP_079076.3
8
989
NM_145910.3
NP_665917.1
8
989
XM_011513167.2
XP_011511469.1
8
1189
XM_011513173.1
XP_011511475.1
6
665
XM_011513174.1
XP_011511476.1
8
935
XM_011513175.1
XP_011511477.1
7
777
XM_011513181.1
XP_011511483.1
4
318
XM_017007205.1
XP_016862694.1
9
1292
XM_017007206.1
XP_016862695.1
9
1210
XM_017007207.1
XP_016862696.1
9
1426
XM_017007208.1
XP_016862697.1
8
1010
XM_017007209.1
XP_016862698.1
8
1219
XM_017007210.1
XP_016862699.1
8
1075
XM_017007211.1
XP_016862700.1
8
1010
XM_017007212.1
XP_016862701.1
8
988
XM_017007213.1
XP_016862702.1
8
988
XM_017007214.1
XP_016862703.1
7
1091
XM_017007215.1
XP_016862704.1
8
988
XM_017007216.1
XP_016862705.1
7
884
XM_017007218.1
XP_016862707.1
10
1193
XM_017007219.1
XP_016862708.1
7
687
XM_017007220.1
XP_016862709.1
8
1010
XM_017007221.1
XP_016862710.1
8
1010
XM_017007222.1
XP_016862711.1
8
1010
XM_017007224.1
XP_016862713.1
9
1054
XM_017007225.1
XP_016862714.1
9
1067
XM_017007226.1
XP_016862715.1
9
1331
XM_017007227.1
XP_016862716.1
7
811
XM_017007228.1
XP_016862717.1
5
426
XM_017007231.1
XP_016862720.1
7
789
XM_017007232.1
XP_016862721.1
8
994
XM_017007233.1
XP_016862722.1
7
744
GenBank mRNA
AB071996.1
-
8
956
AB071997.1
-
8
956
AK027148.1
-
3
182
AK293261.1
-
6
665
AL833472.1
-
7
887
BC028587.2
-
7
1033
4829802
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00380328_m1
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
histone phosphorylation
G2 DNA damage checkpoint
intra-S DNA damage checkpoint
intracellular signal transduction
mitotic cell cycle phase transition
regulation of mitotic cell cycle phase transition
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0