Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128631.2 | NP_001122103.1 | 4 | 614 | ||
NM_001288654.2 | NP_001275583.1 | 4 | 750 | |||
NM_001288655.1 | NP_001275584.1 | 4 | 662 | |||
NM_001321326.1 | NP_001308255.1 | 4 | 560 | |||
NM_024819.6 | NP_079095.3 | 4 | 747 | |||
XM_011525262.2 | XP_011523564.1 | 4 | 779 | |||
XM_017025102.1 | XP_016880591.1 | 5 | 899 | |||
XM_017025105.1 | XP_016880594.1 | 5 | 852 | |||
XM_017025106.1 | XP_016880595.1 | 5 | 794 | |||
GenBank mRNA | AK290755.1 | - | 4 | 563 | ||
BC006472.1 | - | 4 | 636 | 2822306 | ||
BC006546.2 | - | 4 | 636 | 2822306 | ||
BC018132.2 | - | 3 | 508 | 3903964 | ||
BC033299.1 | - | 3 | 647 | 4153431 | ||
BF115235.1 | - | 4 | 623 | |||
BM910634.1 | - | 4 | 661 | |||
DR005747.1 | - | 4 | 662 |