Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195263.1 | NP_001182192.1 | 5 | 831 | ||
NM_024895.4 | NP_079171.1 | 5 | 831 | |||
XM_005270165.2 | XP_005270222.1 | 5 | 806 | |||
XM_011540177.2 | XP_011538479.1 | 6 | 991 | |||
XM_011540178.2 | XP_011538480.1 | 5 | 806 | |||
XM_011540179.2 | XP_011538481.1 | 5 | 806 | |||
XM_011540181.2 | XP_011538483.1 | 5 | 806 | |||
XM_011540182.2 | XP_011538484.1 | 5 | 806 | |||
XM_011540183.2 | XP_011538485.1 | 5 | 806 | |||
XM_017016667.1 | XP_016872156.1 | 5 | 806 | |||
XM_017016668.1 | XP_016872157.1 | 5 | 806 | |||
GenBank mRNA | AK024422.1 | - | 4 | 1096 | ||
AK026862.1 | - | 5 | 796 | |||
AK225136.1 | - | 5 | 803 | |||
BC029054.1 | - | 4 | 756 | 5179973 | ||
FJ617449.1 | - | 6 | 709 |