Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025099.5 | NP_079375.3 | 5 | 805 | ||
XM_006721577.3 | XP_006721640.1 | 5 | 789 | |||
XM_006721578.3 | XP_006721641.1 | 5 | 789 | |||
XM_011524011.2 | XP_011522313.1 | 4 | 550 | |||
GenBank mRNA | AK091077.1 | - | 5 | 756 | ||
BC111783.1 | - | 5 | 756 | 40054070 | ||
DQ451688.1 | - | 5 | 746 | |||
FJ790670.1 | - | 5 | 746 |