Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUFY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040451.2 | NP_001035541.1 | 6 | 930 | ||
NM_001040452.2 | NP_001035542.1 | 7 | 741 | |||
NM_025158.4 | NP_079434.3 | 7 | 916 | |||
XM_005265993.4 | XP_005266050.1 | 4 | 287 | |||
XM_006714922.3 | XP_006714985.1 | 3 | 487 | |||
XM_017009892.1 | XP_016865381.1 | 5 | 713 | |||
XM_017009894.1 | XP_016865383.1 | 7 | 649 | |||
XM_017009895.1 | XP_016865384.1 | 6 | 721 | |||
GenBank mRNA | AB209507.1 | - | 7 | 641 | ||
AF312367.1 | - | 7 | 907 | |||
AF361055.2 | - | 7 | 889 | |||
AK075021.1 | - | 6 | 829 | |||
AK291926.1 | - | 6 | 930 | |||
BC032571.1 | - | 7 | 660 | 5243223 |