Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADAM33 SIRNA Assays ›
Gene Symbol:
ADAM33
Gene Name:
ADAM metallopeptidase domain 33
Gene Aliases:
C20orf153, DJ964F7.1
Chromosome Location:
Chr.20: 3667973 - 3682131 on Build GRCh38
UniGene ID:
Hs.173716
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37240
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ADAM33
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282447.2
NP_001269376.1
7, 8
1356
NM_025220.4
NP_079496.1
7, 8
1356
NM_153202.3
NP_694882.1
7, 8
1356
XM_005260843.1
XP_005260900.1
7, 8
813
XM_006723639.1
XP_006723702.1
7, 8
813
XM_006723640.1
XP_006723703.1
7, 8
813
XM_006723644.2
XP_006723707.1
7, 8
813
XM_011529366.1
XP_011527668.1
7, 8
712
XM_011529367.1
XP_011527669.1
7, 8
673
XM_011529368.1
XP_011527670.1
7, 8
813
XM_011529370.1
XP_011527672.1
7, 8
813
XM_011529371.1
XP_011527673.1
7, 8
813
XM_011529373.1
XP_011527675.1
7, 8
710
XM_017028080.1
XP_016883569.1
7, 8
813
XM_017028081.1
XP_016883570.1
7, 8
734
XM_017028082.1
XP_016883571.1
7, 8
813
XM_017028083.1
XP_016883572.1
7, 8
813
GenBank mRNA
AB055891.1
-
7, 8
757
AF466287.1
-
7, 8
675
AK095953.1
-
6, 7
523
AK123015.1
-
3, 4
1055
AK126487.1
-
4, 5
1831
AK126498.1
-
4, 5
737
AK226148.1
-
5, 6
1279
AK300429.1
-
7, 8
724
AK304037.1
-
7, 8
710
AY223852.1
-
5, 6
526
AY223854.1
-
5, 6
526
AY358314.1
-
7, 8
783
BC125113.1
-
7, 8
717
40118831
Back To Top
More Information
Panther Classification:
Molecular Function -
metalloprotease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
metalloendopeptidase activity
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0