Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317950.1 | NP_001304879.1 | 2 | 324 | ||
NM_030767.5 | NP_110394.3 | 2 | 269 | |||
XM_005252244.2 | XP_005252301.1 | 4 | 441 | |||
XM_005252245.1 | XP_005252302.1 | 3 | 299 | |||
XM_005252247.4 | XP_005252304.1 | 2 | 320 | |||
XM_005252248.2 | XP_005252305.1 | 2 | 437 | |||
XM_006717294.1 | XP_006717357.1 | 2 | 253 | |||
XM_006717295.2 | XP_006717358.1 | 2 | 437 | |||
XM_011519064.2 | XP_011517366.1 | 2 | 437 | |||
XM_011519066.2 | XP_011517368.1 | 2 | 437 | |||
GenBank mRNA | AB075848.1 | - | 2 | 442 | ||
AK126136.1 | - | 2 | 253 | |||
AK131082.2 | - | 2 | 324 | |||
AK160382.1 | - | 2 | 324 | |||
AY703041.1 | - | 3 | 106 | |||
AY703043.1 | - | 1 | 242 | |||
BC055285.1 | - | 2 | 269 | 6143155 |