Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317238.1 | NP_001304167.1 | 12 | 1652 | ||
NM_031921.5 | NP_114127.3 | 14 | 1535 | |||
XM_005244806.3 | XP_005244863.1 | 14 | 1558 | |||
XM_011542243.2 | XP_011540545.1 | 13 | 1271 | |||
XM_011542244.1 | XP_011540546.1 | 14 | 1702 | |||
GenBank mRNA | AB033099.1 | - | 12 | 1741 | ||
AF343078.1 | - | 14 | 1468 | |||
AK024028.1 | - | 12 | 1120 | |||
AK127857.1 | - | 8 | 2306 | |||
AK290586.1 | - | 14 | 1527 | |||
AL834179.1 | - | 4 | 2303 | |||
BC002542.2 | - | 14 | 1461 | 3138578 | ||
BC018701.1 | - | 14 | 1461 | 3139886 |