Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142448.1 | NP_001135920.1 | 5 | 667 | ||
NM_001142449.1 | NP_001135921.1 | 3 | 838 | |||
NM_001142450.1 | NP_001135922.1 | 5 | 817 | |||
NM_001142451.1 | NP_001135923.1 | 4 | 904 | |||
NM_032038.2 | NP_114427.1 | 4 | 904 | |||
XM_006721096.3 | XP_006721159.1 | 5 | 674 | |||
XM_017023757.1 | XP_016879246.1 | 5 | 675 | |||
XM_017023758.1 | XP_016879247.1 | 5 | 671 | |||
XM_017023759.1 | XP_016879248.1 | 5 | 670 | |||
GenBank mRNA | AF212371.1 | - | 4 | 851 | ||
AF370423.1 | - | 5 | 799 | |||
AK095677.1 | - | 4 | 901 | |||
AK289787.1 | - | 4 | 899 | |||
AL390215.1 | - | 3 | 800 | |||
AL524908.3 | - | 5 | 792 | |||
BC006156.1 | - | 4 | 515 | |||
BC008325.1 | - | 4 | 729 | 3501826 | ||
BC038961.1 | - | 5 | 681 | 6053095 | ||
BC047741.1 | - | 4 | 878 | 5757339 | ||
BC065235.1 | - | 5 | 649 | 6161778 |