Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3MBTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007102.2 | NP_001007103.1 | 15 | 1573 | ||
NM_032438.2 | NP_115814.1 | 16 | 1648 | |||
XM_005267161.4 | XP_005267218.1 | 16 | 1824 | |||
XM_006715576.3 | XP_006715639.1 | 16 | 1753 | |||
XM_006715578.3 | XP_006715641.1 | 16 | 1632 | |||
XM_011536179.2 | XP_011534481.1 | 16 | 1631 | |||
XM_011536180.2 | XP_011534482.1 | 16 | 1650 | |||
XM_011536181.2 | XP_011534483.1 | 16 | 1585 | |||
XM_011536183.2 | XP_011534485.1 | 16 | 2181 | |||
XM_011536184.2 | XP_011534486.1 | 16 | 1585 | |||
XM_017011355.1 | XP_016866844.1 | 15 | 1664 | |||
XM_017011356.1 | XP_016866845.1 | 15 | 1556 | |||
GenBank mRNA | AB058701.1 | - | 15 | 1546 | ||
AK092756.1 | - | 6 | 825 | |||
AK094137.1 | - | 16 | 1655 | |||
AK290219.1 | - | 15 | 1565 | |||
BC060845.1 | - | 15 | 1567 | 30342272 |