Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080429.2 | NP_536354.2 | 5 | 582 | ||
XM_011510104.2 | XP_011508406.1 | 5 | 642 | |||
GenBank mRNA | AB066105.1 | - | 5 | 582 | ||
AF159174.1 | - | 5 | 578 | |||
AK314993.1 | - | 5 | 605 | |||
BC069607.1 | - | 5 | 582 | 7262209 | ||
BC074896.2 | - | 5 | 579 | 30915249 | ||
BC074897.2 | - | 5 | 579 | 30915478 |