Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RHOT2 SIRNA Assays ›
Gene Symbol:
RHOT2
Gene Name:
ras homolog family member T2
Gene Aliases:
ARHT2, C16orf39, MIRO-2, MIRO2, RASL
Chromosome Location:
Chr.16: 668083 - 674174 on Build GRCh38
UniGene ID:
Hs.513242
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s40211
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RHOT2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138769.2
NP_620124.1
11
965
XM_005255660.2
XP_005255717.1
11
919
XM_005255661.2
XP_005255718.1
10
865
XM_005255662.2
XP_005255719.1
10
862
XM_005255663.2
XP_005255720.1
10
817
XM_006720970.2
XP_006721033.1
11
902
XM_006720973.2
XP_006721036.1
10
800
XM_017023825.1
XP_016879314.1
11
919
XM_017023826.1
XP_016879315.1
11
916
XM_017023827.1
XP_016879316.1
10
865
XM_017023828.1
XP_016879317.1
10
862
XM_017023829.1
XP_016879318.1
10
817
XM_017023830.1
XP_016879319.1
9
763
XM_017023831.1
XP_016879320.1
11
902
XM_017023832.1
XP_016879321.1
8
663
XM_017023833.1
XP_016879322.1
11
899
XM_017023834.1
XP_016879323.1
9
709
XM_017023835.1
XP_016879324.1
11
914
XM_017023836.1
XP_016879325.1
10
800
XM_017023837.1
XP_016879326.1
11
899
XM_017023838.1
XP_016879327.1
10
780
XM_017023839.1
XP_016879328.1
11
914
XM_017023840.1
XP_016879329.1
10
928
XM_017023841.1
XP_016879330.1
9
1006
XM_017023842.1
XP_016879331.1
10
971
GenBank mRNA
AJ517413.1
-
11
848
AK024450.1
-
7
2395
AK090426.1
-
6
1249
AL834163.1
-
10
983
AY207375.1
-
11
962
BC014942.1
-
11
915
4859240
Back To Top
More Information
Panther Classification:
Molecular Function -
small GTPase
Gene Ontology Categories:
Function(s)
Process(es)
small GTPase mediated signal transduction
cellular homeostasis
mitochondrion transport along microtubule
regulation of small GTPase mediated signal transduction
mitochondrial outer membrane permeabilization
GTPase activity
calcium ion binding
protein binding
GTP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0