Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128225.2 | NP_001121697.1 | 6 | 900 | ||
NM_152264.4 | NP_689477.2 | 6 | 921 | |||
NR_134854.1 | 6 | 990 | ||||
XM_006718381.3 | XP_006718444.1 | 6 | 805 | |||
XM_006718383.3 | XP_006718446.1 | 5 | 697 | |||
XM_006718384.2 | XP_006718447.1 | 6 | 805 | |||
XM_006718385.2 | XP_006718448.1 | 5 | 697 | |||
XM_011520466.1 | XP_011518768.1 | 6 | 805 | |||
XM_011520467.1 | XP_011518769.1 | 6 | 1134 | |||
XM_011520468.2 | XP_011518770.1 | 6 | 1227 | |||
XM_011520469.1 | XP_011518771.1 | 5 | 697 | |||
XM_011520470.1 | XP_011518772.1 | 5 | 671 | |||
XM_017018540.1 | XP_016874029.1 | 6 | 779 | |||
XM_017018541.1 | XP_016874030.1 | 5 | 671 | |||
XM_017018542.1 | XP_016874031.1 | 6 | 779 | |||
GenBank mRNA | AK098651.1 | - | 6 | 990 | ||
BC008853.2 | - | 6 | 766 | 4100291 | ||
BC019016.2 | - | 6 | 785 | 4100057 | ||
BI918365.1 | - | 5 | 482 | |||
BX647491.1 | - | 8 | 1085 | |||
CF132216.1 | - | 5 | 507 |