Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138393.1 | NP_612402.1 | 4 | 560 | ||
GenBank mRNA | AK058112.1 | - | 5 | 535 | ||
AK315744.1 | - | 4 | 535 | |||
AX775999.1 | - | 4 | 560 | |||
AY562244.1 | - | 4 | 456 | |||
BC008201.1 | - | 4 | 560 | 3865853 |