Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178857.5 | NP_849188.4 | 4 | 3951 | ||
GenBank mRNA | AJ491324.2 | - | 3998 | |||
AY168341.1 | - | 4 | 3952 | |||
AY168342.1 | - | 4 | 3952 | |||
AY168343.1 | - | 4 | 3952 | |||
AY168344.1 | - | 3952 | ||||
AY168345.1 | - | 3952 | ||||
AY168346.1 | - | 3952 |