Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308429.1 | NP_001295358.1 | 4 | 964 | ||
NM_138411.2 | NP_612420.1 | 4 | 916 | |||
XM_011526405.2 | XP_011524707.2 | 4 | 621 | |||
XM_011526406.2 | XP_011524708.2 | 4 | 573 | |||
GenBank mRNA | AK125745.1 | - | 4 | 471 | ||
AK131032.1 | - | 4 | 423 | |||
BC004941.1 | - | 4 | 375 | |||
BC012203.1 | - | 4 | 517 | |||
BC071737.1 | - | 4 | 916 | 5192796 |