Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080866.2 | NP_543142.2 | 1 | 364 | ||
XM_017017159.1 | XP_016872648.1 | 1 | 1282 | |||
XM_017017160.1 | XP_016872649.1 | 1 | 96 | |||
GenBank mRNA | AB062418.1 | - | 1 | 197 | ||
AB074812.1 | - | 1 | 345 | |||
AF440402.1 | - | 1 | 332 | |||
AJ295270.1 | - | 1 | 156 | |||
AK074246.1 | - | 1 | 364 | |||
BC022379.1 | - | 1 | 364 | |||
BC126288.1 | - | 1 | 246 | 8992004 |