Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286250.1 | NP_001273179.1 | 8 | 1874 | ||
NM_001286251.1 | NP_001273180.1 | 7 | 1775 | |||
NM_001286254.2 | NP_001273183.1 | 7 | 940 | |||
NM_001323252.1 | NP_001310181.1 | 10 | 2029 | |||
NM_001323257.1 | NP_001310186.1 | 8 | 1766 | |||
NM_001323260.1 | NP_001310189.1 | 9 | 1978 | |||
NM_001323261.1 | NP_001310190.1 | 7 | 1492 | |||
NM_001323263.1 | NP_001310192.1 | 7 | 1496 | |||
NM_001323264.1 | NP_001310193.1 | 6 | 1280 | |||
NM_052904.3 | NP_443136.2 | 9 | 1982 | |||
NR_104421.1 | 7 | 1039 | ||||
NR_136579.1 | 6 | 769 | ||||
NR_136580.1 | 8 | 1255 | ||||
XM_005266813.4 | XP_005266870.1 | 7 | 1387 | |||
XM_017010228.1 | XP_016865717.1 | 6 | 1171 | |||
XM_017010229.1 | XP_016865718.1 | 7 | 1344 | |||
GenBank mRNA | AB067487.1 | - | 7 | 1396 | ||
AF351610.1 | - | 7 | 868 | |||
AK055292.1 | - | 9 | 1868 | |||
AK295479.1 | - | 8 | 1699 | |||
AK297240.1 | - | 7 | 1519 | |||
BC031830.1 | - | 6 | 620 | |||
BC047491.1 | - | 7 | 820 |