Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSGA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304968.1 | NP_001291897.1 | 9 | 1581 | ||
NM_052933.3 | NP_443165.1 | 8 | 1186 | |||
GenBank mRNA | AF294347.1 | - | 8 | 904 | ||
AF431815.1 | - | 8 | 1186 | |||
AK093329.1 | - | 9 | 1580 | |||
BC038415.1 | - | 8 | 1186 | 5172129 |