Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159944.2 | NP_001153416.1 | 9 | 1180 | ||
NM_145245.4 | NP_660288.1 | 10 | 1291 | |||
XM_005272458.4 | XP_005272515.1 | 10 | 1285 | |||
XM_017026239.1 | XP_016881728.1 | 10 | 1277 | |||
GenBank mRNA | AB449874.1 | - | 9 | 1137 | ||
AK091609.1 | - | 4 | 329 | |||
BC014111.2 | - | 10 | 1209 | 4542709 |