Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052958.2 | NP_443190.2 | 12 | 1775 | ||
XM_011517445.2 | XP_011515747.1 | 12 | 1738 | |||
XM_011517446.2 | XP_011515748.1 | 11 | 1696 | |||
XM_011517448.2 | XP_011515750.1 | 11 | 1618 | |||
XM_017013019.1 | XP_016868508.1 | 10 | 1567 | |||
GenBank mRNA | AB056652.1 | - | 11 | 2743 | ||
AK094191.1 | - | 6 | 576 | |||
BC041961.1 | - | 12 | 1775 | 5302156 |