Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018593.4 | NP_061063.2 | 3 | 950 | ||
XM_006715329.2 | XP_006715392.1 | 3 | 1025 | |||
XM_011535422.2 | XP_011533724.1 | 3 | 1025 | |||
XM_017010237.1 | XP_016865726.1 | 2 | 1056 | |||
GenBank mRNA | AB057445.1 | - | 3 | 950 | ||
AK126183.1 | - | 3 | 1025 | |||
AK172789.1 | - | 1 | 171 | |||
BC034031.1 | - | 3 | 1025 | 4604910 | ||
BC066985.1 | - | 3 | 922 | 4827423 |