Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003818.2 | NP_001003818.1 | 8 | 1749 | ||
NM_001198644.1 | NP_001185573.1 | 7 | 1259 | |||
NM_001198645.1 | NP_001185574.1 | 6 | 1163 | |||
NM_058166.4 | NP_477514.1 | 8 | 1550 | |||
GenBank mRNA | AF220030.1 | - | 8 | 1538 | ||
AK023210.1 | - | 5 | 801 | |||
AK027664.1 | - | 8 | 1542 | |||
AK290172.1 | - | 8 | 1749 | |||
AK293295.1 | - | 6 | 992 | |||
AK298301.1 | - | 7 | 1088 | |||
AK316178.1 | - | 8 | 1216 | |||
AK316289.1 | - | 8 | 1344 | |||
BC047564.1 | - | 8 | 1728 | 4825118 | ||
BC065575.1 | - | 8 | 1275 | 4609084 | ||
BG776117.1 | - | 367 | ||||
CR749260.1 | - | 9 | 1445 |