Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153336.2 | NP_699167.2 | 2 | 676 | ||
XM_017015640.1 | XP_016871129.1 | 2 | 302 | |||
XM_017015641.1 | XP_016871130.1 | 2 | 302 | |||
GenBank mRNA | AK094271.1 | - | 1 | 781 | ||
AK127173.1 | - | 2 | 302 | |||
AL833506.1 | - | 1 | 290 | |||
BC035344.1 | - | 2 | 676 | 5185692 |