Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181707.2 | NP_859058.2 | 4 | 483 | ||
XM_005257033.4 | XP_005257090.1 | 4 | 786 | |||
XM_005257034.4 | XP_005257091.1 | 4 | 779 | |||
XM_005257035.4 | XP_005257092.1 | 4 | 420 | |||
XM_005257036.2 | XP_005257093.1 | 4 | 485 | |||
XM_017024166.1 | XP_016879655.1 | 4 | 469 | |||
GenBank mRNA | BC036356.1 | - | 4 | 478 | ||
BC048806.1 | - | 4 | 475 | 5165502 |