Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079810.3 | NP_001073278.1 | 2 | 219 | ||
NM_001199143.1 | NP_001186072.1 | 2 | 219 | |||
NM_152381.5 | NP_689594.4 | 2 | 219 | |||
GenBank mRNA | AK097519.1 | - | 2 | 319 | ||
DB079087.1 | - | 2 | 215 | |||
EF119718.1 | - | 4 | 418 | |||
EF119719.1 | - | 2 | 320 | |||
EF119720.1 | - | 2 | 219 | |||
EF119721.1 | - | 2 | 320 | |||
EF119722.1 | - | 2 | 219 |