Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALS2CR12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127391.2 | NP_001120863.1 | 7 | 955 | ||
NM_001289993.1 | NP_001276922.1 | 7 | 623 | |||
NM_139163.3 | NP_631902.2 | 7 | 955 | |||
XM_011510606.2 | XP_011508908.1 | 7 | 955 | |||
XM_011510610.2 | XP_011508912.1 | 5 | 346 | |||
XM_017003360.1 | XP_016858849.1 | 6 | 538 | |||
XM_017003361.1 | XP_016858850.1 | 6 | 538 | |||
XM_017003362.1 | XP_016858851.1 | 6 | 485 | |||
GenBank mRNA | AB053314.1 | - | 6 | 540 | ||
AK292283.1 | - | 7 | 623 | |||
BC031603.1 | - | 7 | 935 | 5168160 |