Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289002.1 | NP_001275931.1 | 1 | 518 | ||
NM_001289004.1 | NP_001275933.1 | 1 | 518 | |||
NM_001308014.1 | NP_001294943.1 | 1 | 518 | |||
NM_173488.4 | NP_775759.3 | 1 | 518 | |||
XM_005271874.3 | XP_005271931.1 | 1 | 3715 | |||
XM_011543147.2 | XP_011541449.1 | 1 | 3715 | |||
XM_011543148.2 | XP_011541450.1 | 1 | 3715 | |||
XM_011543154.2 | XP_011541456.1 | 1 | 3715 | |||
GenBank mRNA | AF505657.1 | - | 1 | 489 | ||
AK131445.1 | - | 1 | 415 | |||
AY273897.1 | - | 1 | 481 | |||
BC034976.2 | - | 1 | 520 | 4820512 | ||
BG772379.1 | - | 1 | 508 | |||
BM554439.1 | - | 1 | 505 |