Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010940.2 | NP_001010940.1 | 3 | 394 | ||
NM_001308084.1 | NP_001295013.1 | 3 | 394 | |||
NM_001308085.1 | NP_001295014.1 | 2 | 285 | |||
NM_001308086.1 | NP_001295015.1 | 2 | 285 | |||
NR_131756.1 | 2 | 285 | ||||
NR_131757.1 | 2 | 285 | ||||
XM_011518229.2 | XP_011516531.1 | 3 | 395 | |||
XM_011518230.2 | XP_011516532.1 | 3 | 395 | |||
XM_011518231.2 | XP_011516533.1 | 3 | 395 | |||
XM_011518233.2 | XP_011516535.1 | 3 | 297 | |||
XM_011518234.2 | XP_011516536.1 | 2 | 281 | |||
GenBank mRNA | BC136709.1 | - | 3 | 371 | 9020699 | |
BC144373.1 | - | 2 | 262 | |||
BC144374.1 | - | 2 | 262 | |||
BC144375.1 | - | 2 | 262 | |||
BC150564.1 | - | 3 | 372 | 40127624 | ||
BG182005.1 | - | 1 | 64 | |||
CD684431.1 | - | 3 | 422 | |||
CK126031.1 | - | 2 | 303 |