Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001398.2 | NP_001389.2 | 2 | 168 | ||
XM_017026448.1 | XP_016881937.1 | 2 | 118 | |||
GenBank mRNA | AK291860.1 | - | 2 | 103 | ||
AK301211.1 | - | 2 | 100 | |||
BC011792.2 | - | 2 | 85 | 4300082 | ||
BC017408.1 | - | 2 | 109 | 4306900 | ||
BG527600.1 | - | 2 | 169 | |||
U16660.1 | - | 2 | 110 |