Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144666.2 | NP_653267.2 | 42 | 14287 | ||
GenBank mRNA | AB290171.1 | - | 29 | 10987 | ||
AK057314.1 | - | 9 | 2806 | |||
AK128064.1 | - | 10 | 3929 | |||
AL137690.1 | - | 5 | 2251 | |||
AL834222.1 | - | 8 | 3893 | |||
AY358770.1 | - | 6 | 1200 | |||
BC082757.1 | - | 9 | 3530 | |||
BC117301.1 | - | 9 | 2631 | |||
BC117303.1 | - | 9 | 2631 | |||
BC143910.1 | - | 9 | 2601 |