Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010854.1 | NP_001010854.1 | 3 | 457 | ||
NM_001320421.1 | NP_001307350.1 | 3 | 453 | |||
XM_011536497.1 | XP_011534799.1 | 3 | 515 | |||
XM_017021044.1 | XP_016876533.1 | 3 | 421 | |||
XM_017021045.1 | XP_016876534.1 | 3 | 421 | |||
XM_017021046.1 | XP_016876535.1 | 3 | 515 | |||
XM_017021047.1 | XP_016876536.1 | 3 | 254 | |||
GenBank mRNA | AL832848.1 | - | 2 | 135 | ||
BG489583.1 | - | 2 | 123 | |||
BQ426031.1 | - | 372 | ||||
CB988671.1 | - | 484 |