Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008239.2 | NP_001008240.1 | 2 | 813 | ||
NM_145055.3 | NP_659492.1 | 2 | 813 | |||
XM_011525822.1 | XP_011524124.1 | 2 | 664 | |||
XM_017025555.1 | XP_016881044.1 | 2 | 660 | |||
GenBank mRNA | AK289738.1 | - | 2 | 700 | ||
AK292981.1 | - | 2 | 801 | |||
AL832027.2 | - | 1 | 341 | |||
BC016149.1 | - | 1 | 486 | 4040087 | ||
BC084579.1 | - | 2 | 780 | 6141272 | ||
BX640683.1 | - | 1 | 494 |