Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198077.3 | NP_932343.1 | 1 | 263 | ||
NR_024113.1 | 1 | 263 | ||||
GenBank mRNA | AF387611.1 | - | 1 | 224 | ||
AF387612.1 | - | 1 | 224 | |||
AK126929.1 | - | 1 | 263 | |||
AY486331.1 | - | 1 | 233 | |||
AY486332.1 | - | 1 | 233 | |||
BC029538.1 | - | 1 | 233 | 5303467 | ||
DA214065.1 | - | 1 | 263 |