Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136537.1 | NP_001130009.1 | 2 | 442 | ||
XM_006710386.3 | XP_006710449.1 | 2 | 1480 | |||
XM_011540803.2 | XP_011539105.1 | 2 | 1481 | |||
XM_011540805.2 | XP_011539107.1 | 2 | 1478 | |||
XM_017000447.1 | XP_016855936.1 | 2 | 1482 | |||
XM_017000448.1 | XP_016855937.1 | 2 | 1480 | |||
XM_017000449.1 | XP_016855938.1 | 2 | 1481 | |||
GenBank mRNA | AK304611.1 | - | 2 | 361 |
Gene Symbol: PLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209875.1 | - | 16 | 6418 |