Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114938.2 | NP_001108410.2 | 12 | 1840 | ||
NM_001190182.1 | NP_001177111.1 | 12 | 1813 | |||
XM_011540812.1 | XP_011539114.1 | 10 | 2736 | |||
XM_011540814.1 | XP_011539116.1 | 11 | 2667 | |||
XM_017000450.1 | XP_016855939.1 | 10 | 2709 | |||
XM_017000451.1 | XP_016855940.1 | 9 | 2640 | |||
XM_017000452.1 | XP_016855941.1 | 11 | 2637 | |||
XM_017000453.1 | XP_016855942.1 | 9 | 2628 | |||
XM_017000454.1 | XP_016855943.1 | 10 | 1807 | |||
XM_017000455.1 | XP_016855944.1 | 9 | 1354 | |||
XM_017000456.1 | XP_016855945.1 | 9 | 1355 | |||
GenBank mRNA | AK057646.1 | - | 11 | 1743 | ||
AK298104.1 | - | 12 | 1811 | |||
AK303939.1 | - | 12 | 1838 | |||
BC029888.1 | - | 10 | 1463 | 5175485 | ||
BC128177.1 | - | 10 | 1429 | 40113154 |