Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252507.2 | NP_001239436.1 | 2 | 3792 | ||
NM_001282463.1 | NP_001269392.1 | 3 | 3891 | |||
NM_001289139.1 | NP_001276068.1 | 3 | 3885 | |||
XM_017021469.1 | XP_016876958.1 | 2 | 5124 | |||
GenBank mRNA | AK001590.1 | - | 2 | 3735 | ||
AK025915.1 | - | 1 | 1596 | |||
AK074462.1 | - | 1 | 2730 | |||
BC043593.1 | - | 2 | 3775 |