Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197964.4 | NP_932068.2 | 1, 2 | 398 | ||
GenBank mRNA | AF161386.1 | - | 1, 2 | 204 | ||
BC008467.1 | - | 1, 2 | 201 | 4276817 | ||
BI117784.1 | - | 1, 2 | 398 | |||
CB125801.1 | - | 1, 2 | 379 |
Gene Symbol: C7orf55-LUC7L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244584.2 | NP_001231513.1 | 1, 2 | 398 | ||
GenBank mRNA | AK297642.1 | - | 1, 2 | 170 |