Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317906.1 | NP_001304835.1 | 4 | 1026 | ||
NM_001317907.1 | NP_001304836.1 | 5 | 943 | |||
NM_152562.3 | NP_689775.2 | 5 | 943 | |||
XM_006716294.2 | XP_006716357.1 | 5 | 958 | |||
XM_011544415.1 | XP_011542717.1 | 5 | 958 | |||
XM_011544416.2 | XP_011542718.1 | 5 | 652 | |||
XM_011544417.2 | XP_011542719.1 | 5 | 709 | |||
XM_011544418.1 | XP_011542720.1 | 5 | 922 | |||
GenBank mRNA | AK292505.1 | - | 4 | 758 | ||
AK299278.1 | - | 5 | 649 | |||
AL833396.1 | - | 2 | 48 | |||
AL833627.1 | - | 4 | 720 | |||
BC036214.1 | - | 2 | 71 | 5267276 | ||
BM802460.1 | - | 5 | 612 | |||
R95735.1 | - | 2 | 154 |