Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 5 | 527 | ||
XM_006719272.3 | XP_006719335.1 | 5 | 681 | |||
GenBank mRNA | AK127872.1 | - | 5 | 672 | ||
AL832200.1 | - | 1 | 80 | |||
AY727870.2 | - | 5 | 527 | |||
CR749833.1 | - | 4 | 446 | |||
DQ096628.1 | - | 5 | 527 |