Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195811.1 | NP_001182740.1 | 6, 7 | 865 | ||
NM_001195812.1 | NP_001182741.1 | 5, 6 | 703 | |||
NM_001320272.1 | NP_001307201.1 | 7, 8 | 1159 | |||
NM_152666.2 | NP_689879.2 | 7, 8 | 1152 | |||
XM_006711752.2 | XP_006711815.1 | 6, 7 | 1319 | |||
XM_011544115.2 | XP_011542417.1 | 8, 9 | 1227 | |||
XM_011544116.2 | XP_011542418.1 | 8, 9 | 1055 | |||
XM_011544119.2 | XP_011542421.2 | 3, 4 | 1226 | |||
XM_011544120.2 | XP_011542422.1 | 6, 7 | 871 | |||
XM_011544121.2 | XP_011542423.1 | 3, 4 | 360 | |||
XM_011544122.2 | XP_011542424.1 | 3, 4 | 1821 | |||
XM_017000567.1 | XP_016856056.1 | 6, 7 | 1332 | |||
XM_017000568.1 | XP_016856057.1 | 6, 7 | 853 | |||
XM_017000569.1 | XP_016856058.1 | 6, 7 | 2832 | |||
XM_017000570.1 | XP_016856059.1 | 4, 5 | 1572 | |||
GenBank mRNA | AK091691.1 | - | 5, 6 | 677 | ||
AK098092.1 | - | 7, 8 | 1159 | |||
AK295421.1 | - | 6, 7 | 846 | |||
AK307857.1 | - | 6, 7 | 844 | |||
AY391836.1 | - | 5, 6 | 715 | |||
AY461578.1 | - | 6, 7 | 865 | |||
BC101373.1 | - | 6, 7 | 792 | 40025860 | ||
BC101374.1 | - | 6, 7 | 792 | 40025861 | ||
BC101375.1 | - | 6, 7 | 792 | 40025862 |