Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173799.3 | NP_776160.2 | 4 | 1037 | ||
XM_011512538.1 | XP_011510840.1 | 4 | 1022 | |||
XM_017005865.1 | XP_016861354.1 | 3 | 1312 | |||
GenBank mRNA | AK097192.1 | - | 4 | 995 | ||
BC101288.1 | - | 4 | 994 | 40024492 | ||
BC101289.1 | - | 4 | 994 | 40024494 | ||
BC101290.1 | - | 4 | 992 | 40024496 | ||
BC101291.1 | - | 4 | 998 | 40024497 | ||
BX640915.1 | - | 3 | 920 |