Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173799.3 | NP_776160.2 | 1, 2 | 146 | ||
XM_011512538.1 | XP_011510840.1 | 1, 2 | 134 | |||
GenBank mRNA | AK097192.1 | - | 1, 2 | 104 | ||
AL833175.1 | - | 2, 3 | 2886 | |||
BC101288.1 | - | 1, 2 | 103 | 40024492 | ||
BC101289.1 | - | 1, 2 | 103 | 40024494 | ||
BC101290.1 | - | 1, 2 | 103 | 40024496 | ||
BC101291.1 | - | 1, 2 | 103 | 40024497 | ||
EU675310.1 | - | 1, 2 | 146 |