Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000121.3 | NP_000112.1 | 3 | 548 | ||
NR_033663.1 | 3 | 798 | ||||
GenBank mRNA | AK315097.1 | - | 3 | 516 | ||
BC112153.1 | - | 3 | 490 | 8327621 | ||
BC143697.1 | - | 4 | 626 | |||
M34986.1 | - | 3 | 413 | |||
M60459.1 | - | 3 | 518 |