Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317078.1 | NP_001304007.1 | 2 | 419 | ||
NM_153450.2 | NP_703151.2 | 2 | 419 | |||
GenBank mRNA | AY148462.1 | - | 2 | 148 | ||
BC009723.1 | - | 2 | 295 | |||
BC037223.1 | - | 2 | 297 | 5197938 | ||
DB457116.1 | - | 2 | 419 | |||
EB386171.1 | - | 2 | 305 | |||
HE646678.2 | - | 2 | 289 |