Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BEND7 SIRNA Assays ›
Gene Symbol:
BEND7
Gene Name:
BEN domain containing 7
Gene Aliases:
C10orf30
Chromosome Location:
Chr.10: 13438484 - 13528772 on Build GRCh38
UniGene ID:
Hs.498740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s48194
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BEND7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100912.1
NP_001094382.1
4
641
NM_152751.2
NP_689964.2
5
772
XM_005252402.4
XP_005252459.1
5
870
XM_011519390.2
XP_011517692.1
4
563
XM_011519391.2
XP_011517693.1
4
563
XM_011519392.2
XP_011517694.1
5
869
XM_011519393.2
XP_011517695.1
4
658
XM_011519394.2
XP_011517696.1
6
805
XM_011519395.2
XP_011517697.1
4
1165
XM_011519396.2
XP_011517698.1
5
1256
XM_011519397.2
XP_011517699.1
5
811
XM_011519398.2
XP_011517700.1
4
563
XM_011519399.2
XP_011517701.1
4
563
XM_011519400.2
XP_011517702.1
4
563
XM_011519401.2
XP_011517703.1
4
563
XM_011519402.2
XP_011517704.1
4
563
XM_011519403.2
XP_011517705.1
4
563
XM_011519404.2
XP_011517706.1
4
563
XM_011519405.2
XP_011517707.1
4
563
XM_011519406.2
XP_011517708.1
4
563
XM_011519407.1
XP_011517709.1
4
563
XM_011519408.2
XP_011517710.1
4
563
XM_011519410.2
XP_011517712.1
4
563
XM_011519411.2
XP_011517713.1
4
563
XM_011519412.1
XP_011517714.1
4
563
XM_011519413.1
XP_011517715.1
4
563
XM_011519414.1
XP_011517716.1
4
563
XM_017015915.1
XP_016871404.1
6
790
XM_017015916.1
XP_016871405.1
4
563
XM_017015917.1
XP_016871406.1
4
563
XM_017015918.1
XP_016871407.1
5
804
XM_017015919.1
XP_016871408.1
5
826
GenBank mRNA
AK097602.1
-
5
772
BC031618.1
-
4
610
5172600
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-rxbn5:80/10.239.162.255:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0