Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139286.3 | NP_644815.1 | 4 | 544 | ||
XM_017014573.1 | XP_016870062.1 | 4 | 511 | |||
XM_017014574.1 | XP_016870063.1 | 4 | 976 | |||
XM_017014575.1 | XP_016870064.1 | 4 | 506 | |||
GenBank mRNA | AF503918.1 | - | 1 | 185 | ||
BC042534.1 | - | 4 | 499 | 4751173 | ||
BC066300.1 | - | 4 | 529 | 5258904 | ||
BF244254.1 | - | 4 | 542 |