Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178509.5 | NP_848604.3 | 16 | 1684 | ||
XM_005257187.4 | XP_005257244.1 | 16 | 1726 | |||
XM_017024410.1 | XP_016879899.1 | 17 | 1775 | |||
XM_017024411.1 | XP_016879900.1 | 17 | 1767 | |||
XM_017024412.1 | XP_016879901.1 | 16 | 1718 | |||
XM_017024413.1 | XP_016879902.1 | 14 | 1439 | |||
GenBank mRNA | AK122865.1 | - | 16 | 1684 | ||
BC125116.1 | - | 15 | 1607 | 40118892 |