Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM21C SIRNA Assays ›
Gene Symbol:
FAM21C
FAM21EP
Gene Name:
family with sequence similarity 21 member C
family with sequence similarity 21, member A pseudogene
Gene Aliases:
FAM21A, VPEF
-
Chromosome Location:
Chr.10: 45727170 - 45792964 on Build GRCh38
Chromosome Location:
Chr.10: 50021182 - 50067803 on Build GRCh38
UniGene ID:
Hs.365286
Hs.712902
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
s48440
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM21C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001169106.1
NP_001162577.1
18
1790
NM_001169107.1
NP_001162578.1
17
1718
NM_015262.2
NP_056077.2
18
1790
XM_011539566.1
XP_011537868.1
18
1757
XM_011539567.1
XP_011537869.1
17
1685
XM_011539568.1
XP_011537870.1
17
1670
XM_011539569.1
XP_011537871.1
18
1757
XM_011539570.2
XP_011537872.1
17
1646
XM_011539571.1
XP_011537873.1
18
1757
XM_011539572.1
XP_011537874.1
18
1757
XM_017016014.1
XP_016871503.1
18
1757
XM_017016015.1
XP_016871504.1
17
1685
XM_017016016.1
XP_016871505.1
17
1670
XM_017016017.1
XP_016871506.1
16
1598
XM_017016018.1
XP_016871507.1
18
1757
XM_017016019.1
XP_016871508.1
17
1685
XM_017016020.1
XP_016871509.1
17
1685
XM_017016021.1
XP_016871510.1
18
1685
XM_017016022.1
XP_016871511.1
18
1731
XM_017016023.1
XP_016871512.1
17
1603
XM_017016024.1
XP_016871513.1
7
669
XM_017016025.1
XP_016871514.1
8
740
XM_017016026.1
XP_016871515.1
18
1757
GenBank mRNA
AB011164.1
-
18
1771
AK291430.1
-
18
1778
AK293929.1
-
13
1241
AK298385.1
-
16
1455
AK303048.1
-
17
1682
BC053887.1
-
17
1542
BC150611.1
-
18
1755
9056981
Gene Symbol: FAM21EP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_038275.2
5
1694
GenBank mRNA
AK056375.1
-
5
1694
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
retrograde transport, endosome to Golgi
regulation of substrate adhesion-dependent cell spreading
retrograde transport, endosome to plasma membrane
negative regulation of barbed-end actin filament capping
protein binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
phosphatidylinositol-5-phosphate binding
phosphatidylinositol-3-phosphate binding
phosphatidylinositol-3,4-bisphosphate binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0