Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031679.2 | NP_001026849.1 | 7 | 713 | ||
NM_001193460.1 | NP_001180389.1 | 8 | 877 | |||
NM_001193461.1 | NP_001180390.1 | 7 | 575 | |||
NM_198080.3 | NP_932346.1 | 6 | 586 | |||
GenBank mRNA | AK293084.1 | - | 7 | 683 | ||
AK299065.1 | - | 6 | 525 | |||
AK316365.1 | - | 6 | 557 | |||
AY358229.1 | - | 6 | 521 | |||
BC040053.1 | - | 8 | 778 | 6025598 | ||
BX500552.1 | - | 7 | 577 | |||
BX640871.1 | - | 1 | 45 | |||
BX648776.1 | - | 7 | 632 |